Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, ReagentPlus™, ≥99.0%, Solstice

Catalog No. 60044961 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-961 1 L Glass Bottle
60-044-962 2.5 L Glass Bottle
60-044-960 10 L Stainless Steel Can
60-044-963 20 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-044-961 Supplier Solstice Supplier No. 1788101L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent Grade, contains 250 ppm BHT as inhibitor,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.889g/mL at 25°C
Quantity 1 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.407(lit.)
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Boiling Point 65°C
Formula Weight 72.11g/mol
Percent Purity ≥99.0%
Grade ReagentPlus
Chemical Name or Material Tetrahydrofuran, Contains 250 ppm BHT as inhibitor
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H302-H319-H335-H351
DOTInformation 3/PGII
EINECSNumber 203-726-8
RTECSNumber LU5950000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.