Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran (Spectranalyzed™), Fisher Chemical

Catalog No. t4244 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle

Contains no preservatives

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

pH 7 to 8
Melting Point -108.4°C
Absorbance Pass Test
Color Colorless
Density 0.880g/cm³
Boiling Point 66°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.9 %
Linear Formula CH2(CH2)2CH2O
Vapor Pressure 200mbar at 20°C
Viscosity 0.55 cP at 20°C
UN Number 2056
Formula Weight 72.11
Percent Purity ≥99.9%
Grade Spectranalyzed™
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.