Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Spectrophotometric grade, 99.7+%, unstab., packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA41819K2
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L

CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point °C to 108°C
Color Colorless
Appearance Colorless liquid
Density 0.889
Boiling Point 66°C
Quantity 1 L
Flash Point -17°C (1°F)
Odor Ether-like
Physical Form Liquid
Refractive Index 1.407
Assay Percent Range >99.7%
Vapor Pressure 200 hPa (150mm Hg) at 20°C
Beilstein 102391
Solubility Information Miscible with water.
Formula Weight 72.11
Grade Spectrophotometric
Chemical Name or Material Tetrahydrofuran, Unstable
Show More Show Less
Hazard Category H225-H302-H319-H335-H336-H351
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-726-8
RTECSNumber LU5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.