Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Stabilized with BHT, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18607331 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-331 4 x 4 L Amber Glass Bottle
18-607-328 1 L Amber Glass Bottle
18-607-329 4 L Amber Glass Bottle
18-607-330 6 x 1 L Amber Glass Bottle
18-607-332 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-607-331 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP8324LTCS4
Add to Cart
Add to Cart
This item is not returnable. View return policy

HP832, 109-99-9, C4H8O

Spectrum™ Chemical Tetrahydrofuran, Stabilized with BHT, Exceeds A.C.S. Specifications, HPLC Grade , is a colorless, organic liquid with a low viscosity that is water miscible. It has the ability, as an aprotic solvent, to dissolve both polar and non-polar chemical compounds. All HPLC and Ultra-pure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

CAS Max % 1
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.0003
Water 0.0003
Boiling Point 65°C to 66°C
Percent Purity 99.9%
Grade ACS/HPLC
Chemical Name or Material Tetrahydrofuran
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.