Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrahydrofuran, Ultra Low water, No Preservative, BakerDRY™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
Description
- For Use in Organic Synthesis, including Organometallic Synthesis
Specifications
Specifications
Quantity | 1 L |
Flash Point | −14.00°C |
Packaging | Narrow Mouth Amber Glass Bottle, w/ Septum |
Specific Gravity | 1L = 0.88kg |
Formula Weight | 72.11 |
Grade | BakerDRY™ |
Chemical Name or Material | Tetrahydrofuran, Ultra Low water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction