Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl acetate, 98%, Thermo Scientific™

Catalog No. AAB2320530

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 637-64-9
Molecular Formula C7H12O3
MDL Number MFCD00022478
Beilstein 383629
Synonym tetrahydrofurfuryl acetate, 2-acetoxymethyloxolane, tetrahydrofuran-2-yl methyl acetate, 2-furanmethanol, tetrahydro-, acetate, tetrahydro-2-furanmethyl acetate, furfuryl alcohol, tetrahydro-, acetate, 2-acetoxymethyl tetrahydrofuran, tetrahydro-2-furanmethanol acetate, fema no. 3055, 2-furanmethanol, tetrahydro-, 2-acetate
InChI Key AAQDYYFAFXGBFZ-UHFFFAOYSA-N
SMILES CC(=O)OCC1CCCO1
IUPAC Name oxolan-2-ylmethyl acetate
Molecular Weight (g/mol) 144.17
PubChem CID 12506
Formula Weight 144.17
Percent Purity 98%
Density 1.058
Refractive Index 1.437
Chemical Name or Material Tetrahydrofurfuryl acetate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 211-296-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.