Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Acetate 97.0+%, TCI America™
SDP

Catalog No. T010525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T010525G 25 g
T0105500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T010525G Supplier TCI America Supplier No. T010525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 637-64-9
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00022478
InChI Key AAQDYYFAFXGBFZ-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl acetate, 2-acetoxymethyloxolane, tetrahydrofuran-2-yl methyl acetate, 2-furanmethanol, tetrahydro-, acetate, tetrahydro-2-furanmethyl acetate, furfuryl alcohol, tetrahydro-, acetate, 2-acetoxymethyl tetrahydrofuran, tetrahydro-2-furanmethanol acetate, fema no. 3055, 2-furanmethanol, tetrahydro-, 2-acetate
PubChem CID 12506
IUPAC Name oxolan-2-ylmethyl acetate
SMILES CC(=O)OCC1CCCO1

Specifications

Color Colorless
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 195°C
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Acetate
EINECSNumber (5)-0061
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.