Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofurfuryl Propionate 97.0+%, TCI America™
SDP

Catalog No. T101625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101625ML 25 mL
T1016500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T101625ML Supplier TCI America Supplier No. T101625ML

Chemical Identifiers

CAS 637-65-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00047080
InChI Key FMKCDSXOYLTWBR-UHFFFAOYSA-N
Synonym Propionic Acid Tetrahydrofurfuryl Ester
PubChem CID 61183
IUPAC Name oxolan-2-ylmethyl propanoate
SMILES CCC(=O)OCC1CCCO1

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Boiling Point 205°C
Formula Weight 158.20
Percent Purity ≥97.0% (GC)
Chemical Name or Material Tetrahydrofurfuryl Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.