Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Tetrahydrofurfurylamine, 97%

Catalog No. p-7026823
Click to view available options
Quantity:
100 g
500 g

CAS: 4795-29-3 | C5H11NO | 101.15 g/mol

Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrofurfurylamine was used to study specific interactions between unlike molecules by calculating the molar excess enthalpies. It can be used to produce N-tetrahydrofurfuryl-phthalimide with phthalic acid anhydride by heating.

Solubility
Miscible with water.

Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, water/ moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4795-29-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00005373
InChI Key YNOGYQAEJGADFJ-UHFFFAOYNA-N
Synonym tetrahydrofurfurylamine, 2-aminomethyltetrahydrofuran, 2-tetrahydrofurfurylamine, tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro, furfurylamine, tetrahydro, 2-aminomethyl tetrahydrofuran, usaf q-2, tetrahydro-2-furanmethanamine, oxolan-2-yl-methanamine
PubChem CID 253298
IUPAC Name oxolan-2-ylmethanamine
SMILES NCC1CCCO1

Specifications

Density 0.98
Boiling Point 153°C to 154°C
Quantity 100 g
Refractive Index 1.455
UN Number UN2943
Beilstein 102729
Solubility Information Miscible with water.
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material (+/-)-Tetrahydrofurfurylamine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAHYDROFURFURYLAMINE
EINECSNumber 225-351-9
RTECSNumber LV0175000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.