Learn More
Tetraisopropyl methylenediphosphonate, 98%
Description
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Density | 1.08 |
| Boiling Point | 122°C to 124°C (1.5 mmHg) |
| Flash Point | 160°C (320°F) |
| Refractive Index | 1.434 |
| Quantity | 50 g |
| Beilstein | 1080180 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 344.33 |
| Percent Purity | 98% |
| Chemical Name or Material | Tetraisopropyl methylenediphosphonate |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| EINECSNumber | 216-765-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.