Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylamino)silane, 99%

Catalog No. p-7029608
Click to view available options
Quantity:
1 g
10 g

CAS: 1624-01-7 | C8H24N4Si | 204.39 g/mol

Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1624-01-7
Molecular Formula C8H24N4Si
Molecular Weight (g/mol) 204.39
MDL Number MFCD00014859
InChI Key SSCVMVQLICADPI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino silane, silanetetramine, octamethyl, octamethylsilanetetramine, octamethylsilatetramine, silane, tetrakis dimethylamino, silane 48-12 tetrakis, dimethylamine, n,n',n,n'-silanetetrayltetrakis, silanetetramine, n,n,n',n',n,n,n',n'-octamethyl, n,n,n',n',n,n,n',n'-octamethylsilanetetraamine, si nme2 4
PubChem CID 74194
IUPAC Name N-methyl-N-[tris(dimethylamino)silyl]methanamine
SMILES CN(C)[Si](N(C)C)(N(C)C)N(C)C

Specifications

Boiling Point 196°C
Melting Point 15°C
Quantity 1 g
Linear Formula [(CH3)2N]4Si
UN Number UN3129
Flash Point 58°C (136°F)
Solubility Information Reacts with water.
Formula Weight 204.4
Percent Purity 99%
Odor Amine-like
Refractive Index 1.443
Sensitivity Moisture sensitive
Density 0.885
Chemical Name or Material Tetrakis(dimethylamino)silane
Show More Show Less
Hazard Category H226-H261-H302+H312-H314-H335
Hazard Statement GHS H Statement
H261-H226-H314-H318-H302-H312
In contact with water releases flammable gas.
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
EINECSNumber 216-611-2
RTECSNumber VV5455500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.