Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis)
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Color | Yellow to Orange |
| Density | 0.9470g/mL |
| Flash Point | -30°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.99% |
| Packaging | Glass bottle |
| Linear Formula | [(CH3)2N]4Ti |
| Quantity | 25 g |
| Beilstein | 04, IV, 128 |
| Specific Gravity | 0.947 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Reacts violently with water. In contact with water releases flammable gases which may ignite spontaneously. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. Brush off loose parti |
| EINECSNumber | 221-904-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction