Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylamino)titanium(IV), 99.9% (metals basis)

Catalog No. AA4293206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4293206 5 g
AA4293214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4293206 Supplier Thermo Scientific Chemicals Supplier No. 04293206
Add to Cart

CAS: 3275-24-9 | C8H24N4Ti | 224.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3275-24-9
Molecular Formula C8H24N4Ti
Molecular Weight (g/mol) 224.18
MDL Number MFCD00014861
InChI Key MNWRORMXBIWXCI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
PubChem CID 123185
IUPAC Name dimethylazanide;titanium(4+)
SMILES [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C

Specifications

Color Brown-Yellow
Density 0.96 g/mL
Boiling Point 50°C (0.5 mmHg)
Flash Point −30°C (−22°F)
Linear Formula Ti[N(CH3)2]4
Quantity 5 g
UN Number UN3129
Sensitivity Moisture sensitive
Formula Weight 224.19
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetrakis (dimethylamino)titanium (IV)
Show More Show Less
Hazard Category H225-H260-H314-H335
Hazard Statement GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
EINECSNumber 221-904-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.