Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(dimethylsiloxy)silane, 97%

Catalog No. AAB2375414
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 17082-47-2 | C8H24O4Si5 | 324.70 g/mol

It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed for non-volatile NMR standard for high temperature work, in place of the highly volatile and as cross linker for vinyl functional silicones.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17082-47-2
Molecular Formula C8H24O4Si5
Molecular Weight (g/mol) 324.70
MDL Number MFCD00053848
InChI Key UOUILILVWRHZSH-UHFFFAOYSA-N
Synonym tetrakis dimethylsilyl orthosilicate

Specifications

Boiling Point 188°C to 190°C
Melting Point -60°C
Quantity 25 g
Beilstein 1793898
Flash Point 67°C (153°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 328.73
Percent Purity 97%
Refractive Index 1.388
Sensitivity Moisture sensitive
Density 0.884
Chemical Name or Material Tetrakis(dimethylsiloxy)silane
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.