Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(hydroxymethyl)phosphonium chloride, ca. 80% solution in water, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
Glass bottle
124-64-1,7732-18-5
C4H12ClO4P
190.56
MFCD00031687
AKXUUJCMWZFYMV-UHFFFAOYSA-M
tetrakis hydroxymethyl phosphonium chloride,thpc,pyroset tkc,retardol c,tetramethylolphosphonium chloride,tetrakis hydroxymethyl phosphanium chloride,unii-58wb2xcf8i,proban cc,tetrakis hydroxymethyl phosphochloride,ccris 317
31298
tetrakis(hydroxymethyl)phosphanium;chloride
C(O)[P+](CO)(CO)CO.[Cl-]
124-64-1,7732-18-5
C4H12ClO4P
190.56
MFCD00031687
AKXUUJCMWZFYMV-UHFFFAOYSA-M
tetrakis hydroxymethyl phosphonium chloride,thpc,pyroset tkc,retardol c,tetramethylolphosphonium chloride,tetrakis hydroxymethyl phosphanium chloride,unii-58wb2xcf8i,proban cc,tetrakis hydroxymethyl phosphochloride,ccris 317
31298
tetrakis(hydroxymethyl)phosphanium;chloride
C(O)[P+](CO)(CO)CO.[Cl-]
for C-C Bond Formation (Olefination)
Quantity 100mL
Boiling Point 115.0°C
Flash Point 96°C
Color Green-Yellow or Pink
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.3400g/mL
Specific Gravity 1.34
Solubility Information Solubility in water: soluble.
Formula Weight 190.56
Physical Form Solution
Percent Purity 80.0 to 85.0%
Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium chloride
Show More Show Less

GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
Very toxic to aquatic life with lon

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 204-707-7