Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakisepoxy cyclosiloxane

Catalog No. p-7080233
Click to view available options
Quantity:
25 g
100 g

CAS: 257284-60-9 | C28H56O12Si4 | 697.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 257284-60-9
Molecular Formula C28H56O12Si4
Molecular Weight (g/mol) 697.084
MDL Number MFCD22380706
InChI Key BLZDTVROPKIJLZ-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-oxiranylmethoxy propyl cyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-oxiran-2-ylmethoxy propyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, tetrakisepoxy cyclosiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-propyl cyclotetrasiloxane, 2,4,6,8-tetrakis 6,7-epoxy-4-oxaheptyl-2,4,6,8-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis propyl glycidyl ether cyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-glycidyloxy propyl cyclooctanetetrasiloxane, cyclotetrasiloxane,2,4,6,8-tetramethyl-2,4,6,8-tetrakis 3-2-oxiranylmethoxy propyl
PubChem CID 108490
IUPAC Name 2,4,6,8-tetramethyl-2,4,6,8-tetrakis[3-(oxiran-2-ylmethoxy)propyl]-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)CCCOCC2CO2)(C)CCCOCC3CO3)(C)CCCOCC4CO4)CCCOCC5CO5

Specifications

Color Colorless
Physical Form Colorless liquid
Quantity 25 g
Formula Weight 697.09
Chemical Name or Material Tetrakisepoxy cyclosiloxane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.