Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethoxysilane, 98%

Catalog No. AAL0971622
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 681-84-5 | C4H12O4Si | 152.22 g/mol

Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethoxysilane is used in corrosion resistant coatings and in catalysts. It is also used in the production of silicone. Further, it is employed in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers. In addition, it is used in synthetic chemistry to prepare ketals and acetals from ketones and aldehydes respectively.

Solubility
Miscible with ethanol and organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681-84-5
Molecular Formula C4H12O4Si
Molecular Weight (g/mol) 152.22
MDL Number MFCD00008341
InChI Key LFQCEHFDDXELDD-UHFFFAOYSA-N
Synonym tetramethoxysilane, tetramethyl orthosilicate, methyl silicate, tmos, methyl orthosilicate, methyl silicate 39, silane, tetramethoxy, silicic acid, tetramethyl ester, methyl silicate 28, tetramethylorthosilicate
PubChem CID 12682
IUPAC Name tetramethyl silicate
SMILES CO[Si](OC)(OC)OC

Specifications

Boiling Point 121°C to 122°C
Melting Point 4°C to 5°C
Quantity 100 g
UN Number UN2606
Beilstein 1699658
Flash Point 26°C (78°F)
Solubility Information Miscible with ethanol and organic solvents. Immiscible with water.
Formula Weight 152.22
Percent Purity 98%
Odor Ether-like
Refractive Index 1.368
Sensitivity Moisture sensitive
Density 1.032
Chemical Name or Material Tetramethoxysilane
Show More Show Less
Hazard Category H226-H315-H318-H330-H335
Hazard Statement GHS H Statement
H330-H318-H226-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL ORTHOSILICATE
EINECSNumber 211-656-4
RTECSNumber VV9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.