Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl methylenediphosphonate, 98+%

Catalog No. aaa1344106
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 16001-93-7 | C5H14O6P2 | 232.109 g/mol

Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16001-93-7
Molecular Formula C5H14O6P2
Molecular Weight (g/mol) 232.109
MDL Number MFCD00014884
InChI Key XAVFZUKFLWOSOS-UHFFFAOYSA-N
Synonym tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester
PubChem CID 519206
IUPAC Name bis(dimethoxyphosphoryl)methane
SMILES COP(=O)(CP(=O)(OC)OC)OC

Specifications

Boiling Point 152°C to 154°C (2.5 mmHg)
Refractive Index 1.455
Quantity 5 g
Beilstein 1874756
Solubility Information Miscible with water.
Formula Weight 232.11
Percent Purity ≥98%
Chemical Name or Material Tetramethyl methylenediphosphonate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.