Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
64-20-0
C4H12BrN
154.05
MFCD00011626
DDFYFBUWEBINLX-UHFFFAOYSA-M
tetramethylammonium bromide,tetramethylammoniumbromide,tetramethylazanium bromide,methanaminium, n,n,n-trimethyl-, bromide,n,n,n-trimethylmethanaminium bromide,unii-6l9ne9609p,ammonium, tetramethyl-, bromide,methanaminium, n,n,n-trimethyl-, bromide 1:1,c4h12n.br,acmc-209nkm
66137
CHEBI:55317
tetramethylazanium;bromide
C[N+](C)(C)C.[Br-]
64-20-0
C4H12BrN
154.05
MFCD00011626
DDFYFBUWEBINLX-UHFFFAOYSA-M
tetramethylammonium bromide,tetramethylammoniumbromide,tetramethylazanium bromide,methanaminium, n,n,n-trimethyl-, bromide,n,n,n-trimethylmethanaminium bromide,unii-6l9ne9609p,ammonium, tetramethyl-, bromide,methanaminium, n,n,n-trimethyl-, bromide 1:1,c4h12n.br,acmc-209nkm
66137
CHEBI:55317
tetramethylazanium;bromide
C[N+](C)(C)C.[Br-]
Phase transfer catalyst
Quantity 500g
Infrared Spectrum Authentic
Melting Point 230.0°C
Packaging Plastic bottle
Color Gray
Assay Percent Range 98%
Linear Formula (CH3)4NBr
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether
Formula Weight 154.05
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Tetramethylammonium bromide, 98%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fr

GHS Signal Word: Danger

EINECSNumber : 200-581-2

RTECSNumber : BS7600000

TSCA : TSCA