Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium bromide, 98%

Catalog No. p-198315
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 64-20-0 | C4H12BrN | 154.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-20-0
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD00011626
InChI Key DDFYFBUWEBINLX-UHFFFAOYSA-M
Synonym tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm
PubChem CID 66137
ChEBI CHEBI:55317
SMILES [Br-].C[N+](C)(C)C

Specifications

Melting Point 230.0°C
Color Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)4NBr
Quantity 100 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether
IUPAC Name tetramethylazanium;bromide
Formula Weight 154.05
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fr
EINECSNumber 200-581-2
RTECSNumber BS7600000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.