Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
75-57-0
C4H12ClN
109.6
MFCD00011628
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride,tetramethyl ammonium chloride,tetramine chloride,usaf an-8,n,n,n-trimethylmethanaminium chloride,methanaminium, n,n,n-trimethyl-, chloride,unii-dcq9s88703,tetramethylazanium chloride,ammonium, tetramethyl-, chloride,tetramethylammonium chloride tmacl
6379
CHEBI:7070
tetramethylazanium;chloride
C[N+](C)(C)C.[Cl-]
75-57-0
C4H12ClN
109.6
MFCD00011628
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride,tetramethyl ammonium chloride,tetramine chloride,usaf an-8,n,n,n-trimethylmethanaminium chloride,methanaminium, n,n,n-trimethyl-, chloride,unii-dcq9s88703,tetramethylazanium chloride,ammonium, tetramethyl-, chloride,tetramethylammonium chloride tmacl
6379
CHEBI:7070
tetramethylazanium;chloride
C[N+](C)(C)C.[Cl-]
Phase transfer catalyst
Melting Point 420°C
Quantity 250g
Vapor Pressure Negligible
Color White to Ivory
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98+%
Linear Formula (CH3)4NCl
Solubility Information Solubility in water: >60% (20°C). Other solubilities: insoluble in ether,benzene,slightly soluble in ethanol
Formula Weight 109.6
Physical Form Hygroscopic Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material Tetramethylammonium chloride, 98+%
Show More Show Less

GHS P Statement Fatal if swallowed. Toxic in contact with skin. Causes skin irritation. Causes damage to organs if swallowed. Toxic to aquatic life with long lasting effects.

GHS P Statement IF exposed: Call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume

Danger

EINECSNumber : 200-880-8

RTECSNumber : BS7700000

TSCA : TSCA