Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium fluoride tetrahydrate, 98%

Catalog No. AC148070250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 17787-40-5 | C4H12FN·4H2O | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17787-40-5
Molecular Formula C4H12FN·4H2O
Molecular Weight (g/mol) 165.21
MDL Number MFCD00149968
InChI Key HQFTZNVQVRRDLN-UHFFFAOYSA-M
Synonym tetramethylammonium fluoride tetrahydrate, tetramethylazanium fluoride tetrahydrate, methanaminium, n,n,n-trimethyl-, fluoride, tetrahydrate, tetramethylammonium tetrahydrate fluoride, tetramethylammonium ion tetrahydrate fluoride, acmc-1c8r1, tetramethylammoniumfluoridetetrahydrate, methanaminium,n,n,n-trimethyl-,fluoride,tetrahydrate 9ci, tetramethylamine, fluoride, hydrate, hydrate, hydrate, hydrate, methanaminium,n,n,n-trimethyl-, fluoride, tetrahydrate 9ci
PubChem CID 2723808
IUPAC Name tetramethylazanium;fluoride;tetrahydrate
SMILES C[N+](C)(C)C.O.O.O.O.[F-]

Specifications

Melting Point 39°C to 44°C
Flash Point >112°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)4NF·4H2O
Quantity 25 g
Fieser 07,358
Solubility Information Solubility in water: soluble
Formula Weight 165.21
Percent Purity 98%
Chemical Name or Material Tetramethylammonium fluoride tetrahydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.