Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)

Catalog No. AA20932LV
Click to view available options
Quantity:
250 mL
1 L
1 gal.
4 x 1 gal.

CAS: 75-59-2 | C4H13NO | 91.154 g/mol

Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number MFCD00008280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Colorless to Yellow
Density 1.016 g/mL
Boiling Point ∼102°C
Refractive Index 1.384
Linear Formula (CH3)4NOH
Quantity 1 gal.
UN Number UN1835
Beilstein 3558708
Sensitivity Air Sensitive
Merck Index 14,9224
Solubility Information Miscible with water.
Formula Weight 91.15
Percent Purity 99.9999%
Grade Electronic
Concentration or Composition (by Analyte or Components) 25% w/w aq. soln.
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetramethylammonium hydroxide
Show More Show Less
Hazard Category H301-H310-H314-H370-H372
Hazard Statement GHS H Statement
H300-H310-H370-H372-H314
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION
EINECSNumber 200-882-9
RTECSNumber PA0875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.