Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium hydroxide, 25 wt% in methanol, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
75-59-2,67-56-1
C4H13NO
91.154
MFCD00008280
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylammonium hydroxide,tmah,hydroxyde de tetramethylammonium,nmw-w,nmd 3,tetramethyl ammonium hydroxide,unii-5gkp7317q2,ammonium, tetramethyl-, hydroxide,methanaminium, n,n,n-trimethyl-, hydroxide,tetramethylammoniumhydroxide
60966
tetramethylazanium;hydroxide
C[N+](C)(C)C.[OH-]
75-59-2,67-56-1
C4H13NO
91.154
MFCD00008280
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylammonium hydroxide,tmah,hydroxyde de tetramethylammonium,nmw-w,nmd 3,tetramethyl ammonium hydroxide,unii-5gkp7317q2,ammonium, tetramethyl-, hydroxide,methanaminium, n,n,n-trimethyl-, hydroxide,tetramethylammoniumhydroxide
60966
tetramethylazanium;hydroxide
C[N+](C)(C)C.[OH-]
Phase transfer catalyst
Linear Formula (CH3)4NOH
Density 0.9000g/mL
Formula Weight 91.15
Packaging Plastic bottle
Solubility Solubility in water: soluble.
Specific Gravity 0.9
Boiling Point 65.0°C
Melting Point -98.0°C
Quantity 100g
Flash Point 6°C
Chemical Name or Material Tetramethylammonium hydroxide, 25 wt. % in Methanol
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-882-9

RTECSNumber : PA0875000

TSCA : TSCA