Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetramethylammonium Hydroxide, 5-Hydrate, BAKER™, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 1 kg |
| Specific Gravity | 1L = 1.00kg |
| Formula Weight | 181.23 |
| Chemical Name or Material | Tetramethylammonium Hydroxide, 5-Hydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction