Learn More
MP Biomedicals™ Tetramethylammonium Hydroxide Pentahydrate, ≥98%
Shop All MP Biomedicals, Inc ProductsDescription
- Tetramethylammonium Hydroxide Pentahydrate is used in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis, and liquid chromatography.
- TM is also utilized in the synthesis of organic and inorganic compounds such as vinyl halides and tetramethylammonium dithiocarboxylates.
- It is also used in the preparation of iron oxide nanoparticles for application as magnetic resonance imaging contrast agents.
- It is an ion pairing agent that is commonly used in analytical chemistry.
- It is also used as a developer or etchant in semiconductor and photoelectric industries.
Specifications
Specifications
| Item Name | Tetramethylammonium Hydroxide Pentahydrate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Quantity | 10 g |
| Appearance | White |
| Molecular Formula | C4H23NO6 |
| MDL Number | MFCD00149566 |
| InChI Key | MYXKPFMQWULLOH-UHFFFAOYSA-M |
| SMILES | O.O.O.O.O.[OH-].C[N+](C)(C)C |
| IUPAC Name | tetramethylazanium pentahydrate hydroxide |
| Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.