Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium nitrate, 98%, Thermo Scientific™

Catalog No. AA4179030
Click to view available options
:
250g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1941-24-8
Odor Odorless
Molecular Formula C4H12N2O3
Quantity 250g
MDL Number MFCD00043103
UN Number UN1479
Synonym tetramethylammonium nitrate, methanaminium, n,n,n-trimethyl-, nitrate, tetramethylazanium nitrate, methanaminium, n,n,n-trimethyl-, nitrate 1:1, acmc-20ajj7, tetramethylammonium nitrat, tetra methyl ammonium nitrate, ksc174c1t
Solubility Information Soluble in water.
InChI Key QGPVYHSXZFOSRL-UHFFFAOYSA-N
SMILES C[N+](C)(C)C.[N+](=O)([O-])[O-]
IUPAC Name tetramethylazanium;nitrate
Molecular Weight (g/mol) 136.151
PubChem CID 74743
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material Tetramethylammonium nitrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 217-723-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.