Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetramethylcyclopentadiene, mixed isomers, tech. 85%

Click to view available options
Quantity:
1g
5g
4249-10-9
C9H14
122.21
MFCD00145385
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
1,2,3,4-tetramethylcyclopentadiene,tetramethylcyclopentadiene,1,2,3,4-tetramethyl-1,3-cyclopentadiene,tetramethyl cyclopentadiene,tetramethylcyclopentadiene, mixed isomers,pubchem15072,acmc-1an6q,1,3-cyclopentadiene, tetramethyl,2,3,4,5-tetramethylcyclopenta-2,4-diene
138163
1,2,3,4-tetramethylcyclopenta-1,3-diene
CC1=C(C(=C(C1)C)C)C
4249-10-9
C9H14
122.21
MFCD00145385
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
1,2,3,4-tetramethylcyclopentadiene,tetramethylcyclopentadiene,1,2,3,4-tetramethyl-1,3-cyclopentadiene,tetramethyl cyclopentadiene,tetramethylcyclopentadiene, mixed isomers,pubchem15072,acmc-1an6q,1,3-cyclopentadiene, tetramethyl,2,3,4,5-tetramethylcyclopenta-2,4-diene
138163
1,2,3,4-tetramethylcyclopenta-1,3-diene
CC1=C(C(=C(C1)C)C)C
Density 0.808 g/mL
Sensitivity Air and moisture sensitive
Refractive Index 1.472
Flash Point 42°C (108°F)
Quantity 5g
Color Colorless
Boiling Point 55°C to 59°C (13mmHg)
UN Number UN3295
Solubility Information Not miscible mix with mater.
Formula Weight 122.21
Physical Form Liquid
Percent Purity 85%
Grade Technical
Chemical Name or Material Tetramethylcyclopentadiene, Mixed isomers
Show More Show Less

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures