Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene sulfone, 99+%

Catalog No. p-4509948
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name thiolane 1,1-dioxide
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 27.4°C to 27.8°C
Color Yellow
Density 1.2600g/mL
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4820 to 1.4850
Quantity 250 mL
Beilstein 17,I,5
Fieser 01,1144; 02,402; 05,651; 13,256
Merck Index 15,9085
Specific Gravity 1.26
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted mineral acids, soluble in acetone, benzene and toluene, freely soluble in alcohol
Viscosity 10.34 mPa.s (30°C)
Formula Weight 120.17
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material Tetramethylene sulfone
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 204-783-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.