Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene Sulfone, SPEX CertiPrep™
SDP

Catalog No. S3877 Shop All Spex CertiPrep Products
Click to view available options
Quantity:
1 mL

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Tetramethylene sulfone126-33-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 126-33-0, 67-56-1
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name 1λ6-thiolane-1,1-dione
SMILES O=S1(=O)CCCC1

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Chemical Name or Material Tetramethylene sulfone
DOTInformation Nonhazardous
Recommended Storage Ambient
WARNING: Cancer - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.