Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylthiourea 98.0+%, TCI America™
SDP

Catalog No. T0156100G
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 2782-91-4
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00008324
InChI Key MNOILHPDHOHILI-UHFFFAOYSA-N
Synonym tetramethylthiourea, thiourea, tetramethyl, basthioryl, 1,1,3,3-tetramethyl-2-thiourea, tmtu, n,n,n',n'-tetramethylthiourea, urea, thio-, tetramethyl, tetramethyl-2-thiourea, urea, 1,1,3,3-tetramethyl-2-thio, thiourea, n,n,n',n'-tetramethyl
PubChem CID 17725
IUPAC Name 1,1,3,3-tetramethylthiourea
SMILES CN(C)C(=S)N(C)C

Specifications

Melting Point 80°C
Color White
Boiling Point 243°C
Quantity 100 g
Formula Weight 132.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylthiourea
EINECSNumber (2)-2868
RTECSNumber YU2750000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.