Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tetranectin Recombinant Rabbit Monoclonal Antibody (3E5Y7)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535502
Description
Immunogen sequence: GTLGTPQTGS ENDALYEYLR QSVGNEAEIW LGLNDMAAEG TWVDMTGARI AYKNWETEIT AQPDGGKTEN CAVLSGAANG KWFDKRCRDQ LPYICQFGIV.
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
Specifications
Tetranectin | |
Recombinant Monoclonal | |
1.3 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05452, P43025 | |
Clec3b | |
A synthetic peptide corresponding to a sequence within amino acids 103-202 of human CLEC3B/Tetranectin (P05452). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
3E5Y7 | |
Unconjugated | |
Clec3b | |
CLEC3B; C-type lectin domain family 3 member B; C-type lectin domain family 3, member b; LOC100912012; Plasminogen kringle 4-binding protein; tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); tetranectin-like; TN; TNA | |
Rabbit | |
Affinity chromatography | |
RUO | |
100912012, 21922, 7123 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction