Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tetranectin Recombinant Rabbit Monoclonal Antibody (011)
GREENER_CHOICE

Catalog No. PIMA530298
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530298 Supplier Invitrogen™ Supplier No. MA530298
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse Tetranectin/CLEC3B.

Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tetranectin
Applications ELISA
Classification Recombinant Monoclonal
Clone 011
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Clec3b
Gene Accession No. P43025
Gene Alias CLEC3B; C-type lectin domain family 3 member B; C-type lectin domain family 3, member b; LOC100912012; Plasminogen kringle 4-binding protein; tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); tetranectin-like; TN; TNA
Gene Symbols Clec3b
Host Species Rabbit
Immunogen Recombinant Mouse Tetranectin/CLEC3B protein (Met1-Val202).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21922
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.