Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tetranitro blue tetrazolium chloride, 97%

Catalog No. AAJ6108003
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6108003 1 g
AAJ61080MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6108003 Supplier Thermo Scientific Chemicals Supplier No. J6108003
Only null left
Add to Cart

Redox indicator for enzymes | CAS: 1184-43-6 | C40H28Cl2N12O10 | 907.64 g/mol

Redox indicator for enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-43-6
Molecular Formula C40H28Cl2N12O10
Molecular Weight (g/mol) 907.64
MDL Number MFCD00036338
InChI Key VCESGVLABVSDRO-UHFFFAOYSA-L
Synonym TNBT
PubChem CID 129893520
SMILES [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Beilstein 3897475
IUPAC Name 2-[4-[4-[3,5-bis(4-nitrophenyl)tetrazol-2-ium-2-yl]-3-methoxyphenyl]-2-methoxyphenyl]-3,5-bis(4-nitrophenyl)tetrazol-2-ium;dihydrochloride
Formula Weight 999.78
Percent Purity 97%
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material Tetranitro blue tetrazolium chloride
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 214-665-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.