Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylboron sodium, 99.5%

Catalog No. p-199081
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g | CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium;tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Colorless
Loss on Drying 0.5 % max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99.4 % min. (Total base)
Packaging Glass bottle
Linear Formula (C6H5)4BNa
Quantity 25 g
Merck Index 15, 8818
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether
Formula Weight 342.21
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Tetraphenylboron sodium, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
EINECSNumber 205-605-5
RTECSNumber ED3362500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.