Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium bromide, 98+%

Catalog No. AAA1586018
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1586018 50 g
AAA1586009 10 g
AAA1586022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1586018 Supplier Thermo Scientific Chemicals Supplier No. A1586018
Add to Cart

CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol

Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Incompatible with strong oxidizing agents. Stable at room temperature in closed containers under normal storage and handling conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2751-90-8
Molecular Formula C24H20BrP
Molecular Weight (g/mol) 419.30
MDL Number MFCD00011915
InChI Key BRKFQVAOMSWFDU-UHFFFAOYSA-M
Synonym tetraphenylphosphonium bromide, phosphonium, tetraphenyl-, bromide, tetraphenylphosphoniumbromide, tetraphenylphosponium bromide, tetraphenylphosphanium bromide, phosphonium, tetraphenyl-, bromide 1:1, ph4pbr, bromotetraphenylphosphorane, acmc-1ce9k, tetraphenylphosphorus bromide
PubChem CID 2724163
IUPAC Name tetraphenylphosphanium;bromide
SMILES [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 296°C to 302°C
Quantity 50 g
Beilstein 3922383
Sensitivity Hygroscopic
Merck Index 14,9237
Solubility Information Soluble in water.
Formula Weight 419.31
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium bromide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-393-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.