Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylphosphonium iodide, 98+%

Catalog No. AAA1153914
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2065-67-0 | C24H20IP | 466.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2065-67-0
Molecular Formula C24H20IP
Molecular Weight (g/mol) 466.30
MDL Number MFCD00011917
InChI Key AEFPPQGZJFTXDR-UHFFFAOYSA-M
Synonym tetraphenylphosphonium iodide, tetraphenylphosphoniumiodide, tetraphenylphosphanium iodide, acmc-1cpca
PubChem CID 2724164
IUPAC Name tetraphenylphosphanium;iodide
SMILES [I-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 345°C to 350°C
Quantity 25 g
Beilstein 3922216
Sensitivity Light sensitive
Formula Weight 466.31
Percent Purity ≥98%
Chemical Name or Material Tetraphenylphosphonium iodide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-179-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.