Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate, 95%

Catalog No. p-7049876
Click to view available options
Quantity:
250 mg
1 g

CAS: 122451-08-5 | C44H54N4Na4O24S4 | 1243.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122451-08-5
Molecular Formula C44H54N4Na4O24S4
Molecular Weight (g/mol) 1243.119
MDL Number MFCD00151011
InChI Key CDOFYZDFUGMKDJ-UHFFFAOYSA-N
Synonym 4,4',4,4'-porphine-5,10,15,20-tetrayl tetrakis benzenesulfonic acid tetrasodium salt hydrate
PubChem CID 131869494
IUPAC Name sodium;4-[10,15,20-tris(4-sulfophenyl)-21,24-dihydroporphyrin-5-yl]benzenesulfonic acid;dodecahydrate
SMILES C1=CC(=CC=C1C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)S(=O)(=O)O)C8=CC=C(C=C8)S(=O)(=O)O)C9=CC=C(C=C9)S(=O)(=O)O)N3)S(=O)(=O)O.O.O.O.O.O.O.O.O.O.O.O.O.[Na].[Na].[Na].[Na]

Specifications

Quantity 250 mg
Beilstein 5488862
Formula Weight 1239.11 (1022.87 Anhydrous)
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 254-262-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.