Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraspanin-5 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306184MFV610
Description
Tetraspanin-5 Polyclonal antibody specifically detects Tetraspanin-5 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Tetraspanin-5 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
member 8, tetraspanin 5 | |
Produced in rabbits immunized with E. coli-derived Human Tetraspanin-5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
10098 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction