Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraspanin-5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169337 Supplier Novus Biologicals Supplier No. NBP169337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tetraspanin-5 Polyclonal specifically detects Tetraspanin-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Tetraspanin-5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P62079
Gene Alias member 8, tetraspanin 5
Gene Symbols TSPAN5
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TSPAN5(tetraspanin 5) The peptide sequence was selected from the middle region of TSPAN5. Peptide sequence ASRERCGVPFSCCTKDPAEDVINTQCGYDARQKPEVDQQIVIYTKGCVPQ.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10098
Reconstitution Centrifuge lyophilized antibody at 12,000 x g for 20 seconds. Add sterile, distilled water to achieve a final antibody concentration of 1mg/mL (for 100ug samples, add 100uL of dH2O).
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.