Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrathiafulvalene, 99+%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
31366-25-3
C6H4S4
204.34
MFCD00005492
FHCPAXDKURNIOZ-UHFFFAOYSA-N
tetrathiafulvalene,2-1,3-dithiol-2-ylidene-1,3-dithiole,1,3-dithiole, 2-1,3-dithiol-2-ylidene,unii-hy1en16w9t,1,4,5,8-tetrathiafulvalene,ttf,delta-2:2'-bis 1,3-dithiazole,hy1en16w9t,2,2'-bi-1,3-dithiole,delta-2,2'-bi-1,3-dithiole
99451
CHEBI:52444
2-(1,3-dithiol-2-ylidene)-1,3-dithiole
C1=CSC(=C2SC=CS2)S1
31366-25-3
C6H4S4
204.34
MFCD00005492
FHCPAXDKURNIOZ-UHFFFAOYSA-N
tetrathiafulvalene,2-1,3-dithiol-2-ylidene-1,3-dithiole,1,3-dithiole, 2-1,3-dithiol-2-ylidene,unii-hy1en16w9t,1,4,5,8-tetrathiafulvalene,ttf,delta-2:2'-bis 1,3-dithiazole,hy1en16w9t,2,2'-bi-1,3-dithiole,delta-2,2'-bi-1,3-dithiole
99451
CHEBI:52444
2-(1,3-dithiol-2-ylidene)-1,3-dithiole
C1=CSC(=C2SC=CS2)S1
Melting Point 117°C to 120°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Orange
Quantity 250mg
Assay Percent Range 99+%
Merck Index 15, 9390
Formula Weight 204.34
Physical Form Crystals or Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Tetrathiafulvalene
Show More Show Less

EINECSNumber : 250-593-7