Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrolic Acid 98.0+%, TCI America™
SDP

Catalog No. T119925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T119925G 25 g
T11991G 1 g
T11995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T119925G Supplier TCI America Supplier No. T119925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 590-93-2
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00004363
InChI Key LUEHNHVFDCZTGL-UHFFFAOYSA-N
Synonym 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
PubChem CID 68535
IUPAC Name but-2-ynoic acid
SMILES CC#CC(O)=O

Specifications

Melting Point 77°C
Color White-Yellow
Boiling Point 203°C
Quantity 25 g
UN Number 3261
Formula Weight 84.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.