Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEV Cleavage Site Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA1119

Catalog No. PIPA1119


Add to Cart

Description

Description

PA1-119 has been successfully used in Western blot applications. The PA1-119 immunogen is a peptide representing the cleavage site recognized by TEV enzyme.

TEV Cleavage Site Polyclonal antibody specifically detects TEV Cleavage Site in Tag samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TEV Cleavage Site
Polyclonal
Unconjugated
Tobacco Etch Virus
Peptide representing the cleavage site recognized by TEV enzyme
50 μg
Primary
Antibody
IgG
Western Blot
0.5 mg/mL
BBS with no preservative; pH 8.4
Rabbit
Antigen affinity chromatography
RUO
4° C
Liquid
Tag
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit