Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEV Protease Antibody, Novus Biologicals™
SDP

Catalog No. NBP197669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197669 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197669 Supplier Novus Biologicals Supplier No. NBP197669
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEV Protease Polyclonal specifically detects TEV Protease in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TEV Protease
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:1500-1:27000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P04517
Gene Alias P1 Protease, Tobacco Etch Virus Protease
Gene Symbols TEVgp1
Host Species Rabbit
Immunogen This protein-A purified TEV Protease Antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant MBP-and-poly-His-tagged auto inactivation-resistant mutant TEV Protease. (Uniprot: P04517)
Molecular Weight of Antigen 346.16 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Assay by western blot showed no reactivity to MBP.
Target Species Virus
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.