Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEV Tag Monoclonal Antibody (KT88)
GREENER_CHOICE

Catalog No. PIMA549558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549558 Supplier Invitrogen™ Supplier No. MA549558
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This product was used in an Indirect ELISA.

Tobacco Etch Virus (TEV) is a plant virus that primarily infects tobacco plants but can also affect other members of the Solanaceae family. The genome of TEV consists of a single RNA molecule that encodes a polyprotein, which is subsequently cleaved into individual functional proteins by viral proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEV Tag
Applications Western Blot
Classification Monoclonal
Clone KT88
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Alias TEV; Tobacco Etch Virus Protease
Host Species Rat
Immunogen Recombinant TEV tag.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.