Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tex10 Polyclonal Antibody

Catalog No. PI720257
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720257 Supplier Invitrogen™ Supplier No. 720257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Horse, Dog and Cat.

Testis-expressed sequence 10 protein (Tex10) functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex. This complex is recruited to ZNF148 by methylated CHTOP leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes. Tex10 is also involved in chromatin binding, maturation of LSU-rRNA, regulation of DNA-dependent DNA replication initiation, and rRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tex10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.1% BSA and 0.09% sodium azide; pH 7.2-7.4
Gene TEX10
Gene Accession No. Q3URQ0, Q9NXF1
Gene Alias 2610206N19Rik; 2810462N03Rik; bA208F1.2; BC006867; clone 18330; Ipi1; L18; Nbla10363; testis expressed 10; testis expressed gene 10; Testis-expressed protein 10; testis-expressed sequence 10 protein; TEX10
Gene Symbols TEX10
Host Species Rabbit
Immunogen Peptides corresponding to Human TEX10 (aa 240-258, 13-25).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269536, 298065, 54881
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.