Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 173721AP150UL

Description
Testis-expressed sequence 10 protein (Tex10) functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex. This complex is recruited to ZNF148 by methylated CHTOP leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes. Tex10 is also involved in chromatin binding, maturation of LSU-rRNA, regulation of DNA-dependent DNA replication initiation, and rRNA processing.Specifications
TEX10 | |
Polyclonal | |
Unconjugated | |
TEX10 | |
Nbla10363, testis expressed 10, TEX10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269536, 298065, 54881 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3URQ0, Q9NXF1 | |
TEX10 | |
TEX10 Fusion Protein Ag11014 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction