Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEX101 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $610.00
Specifications
Antigen | TEX101 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TEX101 Polyclonal specifically detects TEX101 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TEX101 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
83639 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ELYCQKGLSMTVEADPANMFNWTTEEVETCDKGALCQETILIIKAGTETAILATKGCIPEGEEAITIVQHSSPPG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
cancer/testis antigen 131, Cell surface receptor NYD-SP8, CT131, MGC4766, NYD-SP8, PRO1884, Scleroderma-associated autoantigen, SGRGGTPR867, Spermatogenesis-related gene protein, TES101RP, testis expressed 101, testis expressed sequence 101, testis-expressed protein 101, testis-specific protein TES101RP | |
TEX101 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title