Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TEX101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120611 Supplier Invitrogen™ Supplier No. PA5120611
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549. Immunogen sequence: LYCQKGLSMT VEADPANMFN WTTEEVETCD KGALCQETIL IIKAGTETAI LATKGCIPEG EEAITIVQHS SPPGLIVTSY SNYCEDSFCN DKDSLSQFWE FSETTAS.

May play a role in signal transduction and promote protein tyrosine phosphorylation. Target protein is a glycoprotein whose aggregation induced a rapid increase in tyrosine phosphorylation of Syk kinase and LAT adaptor, calcium flux, and release of secretory components.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEX101
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TEX101
Gene Accession No. Q924B5, Q9BY14, Q9JMI7
Gene Alias cancer/testis antigen 131; cell surface receptor NYD-SP8; CT131; GTPR867; NYD-SP8; PRO1884; Scleroderma-associated autoantigen; SGRG; SPATA44; spermatogenesis associated 44; spermatogenesis-related gene protein; TES101RP; testis expressed 101; testis expressed sequence 101; testis-expressed protein 101; testis-expressed sequence 101 protein; testis-specific protein TES101RP; TEX101; UNQ867/PRO1884
Gene Symbols TEX101
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 44-150 of human TEX101 (NP_113639.4).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207113, 56746, 83639
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.