Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEX101 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549381
Description
May play a role in signal transduction and promote protein tyrosine phosphorylation.
Specifications
TEX101 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9BY14 | |
TEX101 | |
KLH conjugated synthetic peptide between 214-246 amino acids from the C-terminal region of human TEX101. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TEX101 | |
Testis-expressed sequence 101 protein, Cell surface receptor NYD-SP8, Scleroderma-associated autoantigen, Spermatogenesis-related gene protein, TEX101, SGRG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83639 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction